1,4-Dithioerythrit ≥99%, weißes kristallines Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
Dithioerythritol is an erythro isomer of 2,3-dihydroxy-1,4-dithiolbutane, and an isomer of 1,4-dithiothreitol which is a reagent for maintaining thiols in the reduced state; prevents oxidation of sulfhydryl-containing proteins during SDS-polyacrylamide gel electrophoresis. It also quantitatively reduces disulfides.
Dithioerythritol is used to prepare a matrix for fast-atom bombardment mass spectroscopy. It has been reported to be used in 2-D electrophoretic analysis of proteins and immunoglobulins, mitochondrial studies to block the opening of the transition pore and to study permeability transition, glial and neuronal glycine transporters have been investigated, utilised in a mass spectrometric mapping study of disulphide bonds in recombinant human interleukin-13.
Soluble in water (50 mg/ml - clear to slightly hazy, colourless solution).
Bitte beachten Store at +4 °C, protect from light, store under nitrogen
Formel:
C₄H₁₀O₂S₂ Molekulargewicht: 154.25 g/mol Siedepunkt: 286 Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00063750 CAS-Nummer: 6892-68-8 EINECS: 229-998-8 UN-Nummer: 3335 ADR: 3 III |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Reference |
Melting Point | 80 - 90 °C |
UV/ VIS: OD283 (0,02 M aq soln) | ≤0,04 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...