Magnesiumchlorid Hexahydrat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
Synonyme:
Magnesium dichloride
10797.14
10797.22
10797.14EA
230
EUR
10797.14
10797.22
Magnesiumchlorid Hexahydrat ≥99,999% (Metall-Basis), Puratronic®
Magnesiumchlorid Hexahydrat
Purity: 99.999%
CAS No.: 7791-18-6
Molecular Formula: MgCl2.6H2O
Formula Weight: 203.30 (95.22anhy)
Physical Form: Crystalline Aggregates
Density: 1.570
Appearance: White
Odour: Odourless
Melting Point: 117° dec.
Merck Reference: 14,5662
MDL No.: MFCD00149781
Formel:
MgCl₂·6H₂O Siedepunkt: 1412 °C (1013 hPa) Schmelzpunkt: 117 °C (dec.) Dichte: 1.570 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149781 CAS-Nummer: 7791-18-6 EINECS: 232-094-6 Merck Index: 14,5662 |