Natriumselenit Pentahydrat ≥99,0% (RT), Puriss. p.a. analytical reagent, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

00163-100G 00163-25G
00163-100GEA 121.11 EUR
00163-100G 00163-25G
Natriumselenit Pentahydrat ≥99,0% (RT), Puriss. p.a. analytical reagent, Fluka™
Natriumselenit Pentahydrat
Formel: Na₂O₃Se·5H₂O
Schmelzpunkt: Dec.
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149173
CAS-Nummer: 26970-82-1
EINECS: 233-267-9
UN-Nummer: 2630
ADR: 6.1,I

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Calcium (Ca) max. 20 ppm
Cadmium (Cd) max. 5 ppm
Cobalt (Co) max. 5 ppm
Chromium (Cr) max. 5 ppm
Copper (Cu) max. 5 ppm
Iron (Fe) max. 10 ppm
Potassium (K) max. 50 ppm
Magnesium (Mg) max. 5 ppm
Manganese (Mn) max. 5 ppm
Nickel (Ni) max. 5 ppm
Lead (Pb) max. 5 ppm
Zinc (Zn) max. 5 ppm
Chloride (Cl) max. 50 ppm
Total N max. 50 ppm
Selenate and sulfate (as SO₄) max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR