tetra-Natriumdiphosphat Decahydrat 99,0-103,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
Purity: 99.0-103.0%
CAS No.: 13472-36-1
Molecular Formula: Na4O7P2.10H2O
Formula Weight: 446.06 (265.90anhy)
Physical Form: Crystalline
Density: 1.82
pH: 9.5-10.5
Melting Point: 79.5°
Merck Reference: 14,9240
MDL No.: MFCD00149200
Formel:
Na₄P₂O₇·10H₂O Molekulargewicht: 446.06 g/mol Schmelzpunkt: 79.5 °C Dichte: 1.82 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149200 CAS-Nummer: 13472-36-1 EINECS: 231-767-1 Merck Index: 14,9240 |