Natriumiodid Dihydrat ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium iodide hydrate

10871.06 10871.14 10871.22
10871.06EA 51.5 EUR
10871.06 10871.14 10871.22
Natriumiodid Dihydrat ≥99,999% (Metall-Basis), Puratronic®
Natriumiodid Dihydrat

Purity: 99.999% Molecular Formula: NaI.xH2O Formula Weight: 149.89(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic

Formel: INa·2H₂O
Schmelzpunkt: 64.3 °C
Dichte: 2.67 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149211
CAS-Nummer: 13517-06-1
EINECS: 231-679-3
UN-Nummer: 3077
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR