Natriumdichromat Dihydrat ≥99%, Reagenzqualität

Lieferant: Thermo Fisher Scientific
Gefahr


Packaged under Argon in resealable ChemSeal™ bottles.

12611.22 12611.36 12611.A3
12611.22EA 31.6 EUR
12611.22 12611.36 12611.A3
Natriumdichromat Dihydrat ≥99%, Reagenzqualität
Natriumdichromat Dihydrat

CAS No.: 7789-12-0 Molecular Formula: Cr2Na2O7.2H2O Formula Weight: 298.00 (261.97anhy) Physical Form: Crystalline Density: 2.348 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: 100° -2H2O Solubility: Soluble in water Merck Reference: 14,8609 UN No.: 3087 MDL No.: MFCD00149166

Formel: Na₂Cr₂O₇·2H₂O
Schmelzpunkt: 100 °C (-2H₂O)
Dichte: 2.348
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149166
CAS-Nummer: 7789-12-0
EINECS: 234-190-3
UN-Nummer: 3087
ADR: 5.1,II
Merck Index: 14,8609

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR