(±)-1,2,4-Butantriol 96%
Lieferant: Thermo Fisher Scientific
L05138.06
L05138.14
L05138.06EA
27.6
EUR
L05138.06
L05138.14
(±)-1,2,4-Butantriol 96%
(±)-1,2,4-Butantriol
Purity: 96%
CAS No.: 3068-00-6
Molecular Formula: C4H10O3
Formula Weight: 106.12
Flash Point: 188°(370°F)
Refractive Index: 1.4750
Melting Point: -20°
Boiling Point: 190-191°/18mm
Beilstein Reference: 1733456
MDL No.: MFCD00002929
Formel:
C₄H₁₀O₃ Siedepunkt: 190…191 °C (18 mmHg) Schmelzpunkt: –20 °C Dichte: 1.182 Flammpunkt: 188 °C (370 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002929 CAS-Nummer: 3068-00-6 EINECS: 221-323-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...