Kupfer ≥99,999% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)
Lieferant: Thermo Fisher Scientific
11378.FG
11378.FY
11378.HL
11378.FGEA
154
EUR
11378.FG
11378.FY
11378.HL
Kupfer ≥99,999% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)
Copper
Purity: 99.999%
CAS No.: 7440-50-8
Physical Form: ≈11.2g/25x50mm
Odour: Odourless
MDL No.: MFCD00010965
Formel:
Cu Siedepunkt: 2595 °C (1013 hPa) Schmelzpunkt: 1083 °C Dichte: 8.94 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010965 CAS-Nummer: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...