Iod ≥99,5%, kristallin
Lieferant: Thermo Fisher Scientific
|
Achtung
|
14248.14
14248.30
14248.A1
14248.14EA
54.7
EUR
14248.14
14248.30
14248.A1
Iod ≥99,5%, kristallin
Iod
Purity: 99.5%
CAS No.: 7553-56-2
Molecular Formula: I2
Formula Weight: 253.81
Physical Form: Solid
Density: 4.930
Melting Point: 183-186° subl.
UN No.: 3495
MDL No.: MFCD00011355
Formel:
I₂ Molekulargewicht: 253.81 g/mol Siedepunkt: 184.3 Schmelzpunkt: 183…186 °C (Subl.) Dichte: 4.930 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011355 CAS-Nummer: 7553-56-2 EINECS: 231-442-4 UN-Nummer: 3495 ADR: 8 III Merck Index: 13,05036 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...