CAPS (3-(Cyclohexylamino)propansulfonsäure), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Cyclohexylaminopropanesulphonic acid

M-Clarity™ quality level = MQ200

8.43280.0025 8.43280.0100
8.43280.0025EA 59.5 EUR
8.43280.0025 8.43280.0100
CAPS (3-(Cyclohexylamino)propansulfonsäure), Sigma-Aldrich®
CAPS (3-(Cyclohexylamino)propansulfonsäure)
Formel: C₆H₁₁NH(CH₂)₃SO₃H
Schmelzpunkt: 324 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003837
CAS-Nummer: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR