ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na), Calbiochem®
Lieferant: Merck Millipore Calbiochem
Synonyme: NADP monosodium salt, NADP Sodium, Nadide phosphate monosodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl hydrogen phosphate, Nicotinamide-adenine dinucleotide phosphate monosodium salt, NADP sodium salt, NADP-Na
481971-1 481971-5 481971-500
481971-1EA 479 EUR
481971-1 481971-5 481971-500
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na), Calbiochem®
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form(NADP-Na)
Formel: C₂₁H₂₇N₇NaO₁₇P₃
Molekulargewicht: 765.39 g/mol
MDL Number: MFCD00036973
CAS-Nummer: 1184-16-3
EINECS: 214-664-6