ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na)
Lieferant: Thermo Fisher Scientific
Synonyme:
NADP monosodium salt, NADP Sodium, Nadide phosphate monosodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl hydrogen phosphate, Nicotinamide-adenine dinucleotide phosphate monosodium salt, NADP sodium salt, NADP-Na
CAS No.: 1184-16-3
Molecular Formula: C21H27N7NaO17P3
Formula Weight: 765.40
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4779954
MDL No.: MFCD00036973
Formel:
C₂₁H₂₇N₇NaO₁₇P₃ Molekulargewicht: 765.39 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00036973 CAS-Nummer: 1184-16-3 EINECS: 214-664-6 |