Kupfer(II)phthalocyanin ∼95% (dye-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15

269150250 269152500
ACRO269150250EA 43.8 EUR
ACRO269150250 ACRO269152500
Kupfer(II)phthalocyanin ∼95% (dye-Basis)
Copper(II) phthalocyanine
Formel: C₃₂H₁₆CuN₈
Schmelzpunkt: 480 °C
MDL Number: MFCD00010719
CAS-Nummer: 147-14-8
EINECS: 205-685-1
Merck Index: 12,02584

Bestellen

Spezifikation Testergebnisse

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Titration with Na2S2O3 ≥94 % (ex Cu)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR