(-)-Chinin Hydrochlorid Dihydrat, weißes Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate
Quinine Hydrochloride is an alkaloid originally isolated from the Cinchona family of South American trees. It is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent.
Quinine is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent; also used as a resolving agent. A potassium channel blocker. Also used as a flavoring agent in carbonated beverages, a skeletal muscle relaxant and an antiprotozoal agent for fish. A chiral catalyst used in highly enantioselective [2+2] cycloadditions. A resolving agent for carboxylic acids. Catalyzes the kinetic resolution of furanones.
Potassium channel blocker.
Soluble in water (1 g/16 ml at 25 °C; 1 g/0,5 ml at 100 °C), ethanol (1 g/1 ml), glycerol (~1 g/7 ml), chloroform (~1 g/ml), or ether (1 g/350 ml).
Formel:
C₂₀H₂₅ClN₂O₂·2H₂O Schmelzpunkt: 115…116 °C (lit.) Flammpunkt: 122 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151248 CAS-Nummer: 6119-47-7 EINECS: 205-001-1 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...