(-)-Chinin Hydrochlorid Dihydrat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate
M-Clarity™ quality level = MQ300
W297607-100G-K
W297607-1KG-K
W297607-5KG-K
SIALW297607-100G-KEA
69.5
EUR
SIALW297607-100G-K
SIALW297607-1KG-K
SIALW297607-5KG-K
(-)-Chinin Hydrochlorid Dihydrat, Sigma-Aldrich®
(-)-Chinin Hydrochlorid Dihydrat
Formel:
C₂₀H₂₅ClN₂O₂·2H₂O Schmelzpunkt: 158…160 °C Flammpunkt: 122 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151248 CAS-Nummer: 6119-47-7 EINECS: 205-001-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...