(-)-Chinin, wasserfrei 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
163700100
163700500
163702500
ACRO163700100EA
55
EUR
ACRO163700100
ACRO163700500
ACRO163702500
(-)-Chinin, wasserfrei 99%
(-)-Chinin
Formel:
C₂₀H₂₄N₂O₂ Schmelzpunkt: 176…177 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00198096 CAS-Nummer: 130-95-0 EINECS: 205-003-2 Merck Index: 12,08245 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 174 to 178 °C |
Loss on drying | ≤1 % (105 °C, 2 hrs) |
Sulfated ash | ≤0.1 % |
Specific optical rotation | -159° to -172° (20 °C, 589 nm) (c=1, C2H5OH) (on dried substance) |
Titration with HClO4 | ≥98.5 % (On dried substance) |
Chloride (Cl) | ≤250 ppm |
Sulfate (SO4) | ≤500 ppm |
Copper (Cu) | ≤50 ppm |
Iron (Fe) | ≤50 ppm |
Lead (Pb) | ≤50 ppm |
Nickel (Ni) | ≤50 ppm |
Potassium (K) | ≤100 ppm |
Zinc (Zn) | ≤50 ppm |
Insoluble matter | ≤0.01 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...