Halbmikro- und Analysenwaagen, Excellence Plus XP-Serie
Lieferant: Mettler - Toledo
11106027 11106024 11106030 11106032 11106036
611-2015EA 0 EUR
611-2015 611-2013 611-2017 611-2018 611-2019
Halbmikro- und Analysenwaagen, Excellence Plus XP-Serie
Waagen
The XP analytical balances provide high performance weighing with improved linearity safety and security. This is achieved with the introduction of QM toolbox, SmartSens and MinWeigh functions alongside the SmartGrid technology, which provide fast and accurate weighing. This range is therefore particularly suited where regulatory compliance is required. The draught shield and drip tray can be completely, quickly and easily removed as before but now SmartSens allows the operator to weigh without touching the balance, thereby smoothing the work-flow and reducing spillage and contamination risks. MinWeigh warns the user if the minimum weight requirement, as laid down in USP or local GMP procedures, has been violated. The design also features a large, colour, backlit touch screen that facilitates direct access to all functions and an extended range of applications that include density determinations, differential weighing formulation, target weighing and statistical evaluation.
  • SmartGrid and ErgoClips for fast results and secure sample handling of awkward shaped vessels
  • SmartSens can operate the motorised doors without touching the balance
  • QM toolbox includes user administration and password protection
  • Level Control warns the user if the balance is inclined and guides the user how to re-level the unit
  • Improved connectivity which include Ethernet and Bluetooth options
Balances with solid pan
Four place balances without SmartGrid or motorised draught shield doors. These balances feature MonoBloc HighSpeed technology with pan supported from below, but with the other XP features.
Four place balances without SmartGrid or motorised draught shield doors. These balances feature MonoBloc HighSpeed technology with pan supported from below, but with the other XP features.
Bestellinformation: Ebenfalls sind OIML-zertifizierte Versionen für alle unten aufgeführten Modelle sowie eine vollständige Palette an Zubehör für Excellence-Waagen erhältlich – besuchen Sie bitte die Avantor-Website oder wenden Sie sich an Ihre örtliche Avantor-Niederlassung.