Gold standard syringes for autosampler
Lieferant: Agilent
5181-1267 5181-1273 5181-3321 5181-3356 5181-3360 5181-3361 5181-8806 5181-8809 5181-8810 5181-8811 5181-8813 5182-0830 5182-0834 5182-0835 5182-0875 5183-0314 5183-0316 5183-0318 5183-2042 5183-2058 5183-4728 5183-4729 5183-4730 9301-0713 9301-0714 9301-0725 9301-0891 9301-0892 5188-5246 5188-5247
548-0916EA 73.3 EUR
548-0916 548-0917 HEWL5181-3321 HEWL5181-3356 HEWL5181-3360 HEWL5181-3361 HEWL5181-8806 548-0919 HEWL5181-8810 HEWL5181-8811 HEWL5181-8813 HEWL5182-0830 HEWL5182-0834 HEWL5182-0835 HEWL5182-0875 HEWL5183-0314 548-0925 HEWL5183-0318 HEWL5183-2042 HEWL5183-2058 HEWL5183-4728 HEWL5183-4729 HEWL5183-4730 548-0926 548-0927 HEWL9301-0725 548-0928 548-0929 HEWL5188-5246 HEWL5188-5247
Gold standard syringes for autosampler
Spritzen Spritzen für die Chromatographie
These syringes with fixed needle are compatible with Agilent autosamplers 7693A and 7683.
* with PTFE-tipped plunger