Desiccators
Lieferant: Bel-Art Products, a Part of SP
F420300000 F420310000 F420320000
467-0075EA 131 EUR
467-0075 467-0076 467-0077
Desiccators
Exsikkatoren Exsikkatoren, Kunststoff
Desiccators with white PP base and clear PC domed lid with large ribbed knob. Desiccant can be placed on or under the PP plate.
  • Space saving, maximum interior clearance and volume with a minimal footprint
  • Neoprene O-ring ensures tight seal down to 0 °C without grease, and will not be harmed if grease is accidentally applied
Bestellinformation: Supplied with PP plate which is 3.2 mm thick with 3.2 mm diameter holes for air circulation.