1,6-Diphenyl-1,3,5-hexatrien, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DPH, 1,6-Diphenylhexa-1,3,5-triene, (6-Phenylhexa-1,3,5-trien-1-yl)benzene

M-Clarity™ quality level = MQ200

D208000-1G D208000-5G
SIALD208000-1GEA 71.1 EUR
SIALD208000-1G SIALD208000-5G
1,6-Diphenyl-1,3,5-hexatrien, Sigma-Aldrich®
1,6-Diphenyl-1,3,5-hexatrien

Used an alternative to Nile red for fluorescent detection of lipid droplets

Formel: (C₆H₅)CHCHCHCHCHCH(C₆H₅)
Siedepunkt: 351 °C (1013 hPa)
Schmelzpunkt: 204…205 °C
Dichte: 1.075 g/cm³ (20 °C)
MDL Number: MFCD00004793
CAS-Nummer: 1720-32-7
EINECS: 217-011-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR