3-Amino-1H-1,2,4-triazol 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Amino-1,2,4-triazole, Amitrole, 3-Amino-1H-1,2,4-triazole

A14871.0B A14871.18 A14871.30
A14871.0BEA 292 EUR
A14871.0B A14871.18 A14871.30
3-Amino-1H-1,2,4-triazol 96%
3-Amino-1H-1,2,4-triazol

Purity: 96% CAS No.: 61-82-5 Molecular Formula: C2H4N4 Formula Weight: 84.08 Physical Form: Powder Density: 1.138 Appearance: White to cream Odour: Odourless Melting Point: 147-153° Beilstein Reference: 107687 Merck Reference: 14,489 UN No.: 3077 MDL No.: MFCD00005230

Formel: C₂H₄N₄
Siedepunkt: 347 °C (1013 hPa)
Schmelzpunkt: 147…153 °C
Dichte: 1.138
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00053362
CAS-Nummer: 61-82-5
EINECS: 200-521-5
ADR: 9,III
Merck Index: 14,489

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR