Chinaldinrot, rein
Lieferant: Thermo Fisher Scientific
Synonyme:
2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide
377510050
377510250
377511000
ACRO377510050EA
70.8
EUR
ACRO377510050
ACRO377510250
ACRO377511000
Chinaldinrot, rein
Chinaldinrot
Formel:
C₂₁H₂₃IN₂ Molekulargewicht: 430.33 g/mol Schmelzpunkt: 240 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011968 CAS-Nummer: 117-92-0 EINECS: 204-221-5 Merck Index: 12,08230 |
Spezifikation Testergebnisse
Appearance | Grey to green-black to dark blue Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...