3,5-Dichlor-4-fluorbenzotrifluorid
Lieferant: Thermo Scientific
Synonyme:
1,3-Dichloro-2-fluoro-5-(trifluoromethyl)benzene, 3,5-Dichloro-4-fluorobenzotrifluoride
SEW02795.1
SEW02795.10
MAYBSEW02795.1EA
0
EUR
MAYBSEW02795.1
MAYBSEW02795.10
3,5-Dichlor-4-fluorbenzotrifluorid
3,5-Dichlor-4-fluorbenzotrifluorid
Formel:
C₇H₂Cl₂F₄ Siedepunkt: 166…168 °C (1013 hPa) Dichte: 1,55 g/cm³ (20 °C) |
MDL Number:
MFCD00068183 CAS-Nummer: 77227-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...