HyFlex® 11-591 Industrial Gloves
Lieferant: Ansell
11-591/10 11-591/11 11-591/12 11-591/6 11-591/7 11-591/8 11-591/9
ANSE11-591/10EA 39.1 EUR
ANSE11-591/10 ANSE11-591/11 ANSE11-591/12 ANSE11-591/6 ANSE11-591/7 ANSE11-591/8 ANSE11-591/9
HyFlex® 11-591 Industrial Gloves
Handschuhe
HyFlex® 11-591 industrial gloves provide enhanced comfort, with a 20% lighter design that allows extended use on varied tasks.
  • Ultra-lightweight design
  • 12 times greater cut resistance for reliable hand protection
  • Palm dipped, thumbcrotch reinforcement
  • Silicone-free and anti-static
HyFlex® 11-591 industrial gloves offer 12 times greater cut resistance thanks to the Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Featuring a 20% lighter design, these gloves provide enhanced comfort, and allow workers to perform varied tasks while keeping hands protected throughout extended use. Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting handling in abrasive conditions. EN 407 contact heat level 1 to protect hands while handling hot parts.
HyFlex® 11-591 industrial gloves are recommended for use in automotive, glass manufacturing, heavy manufacturing, metal fabrication, machinery, equipment, and pulp/paper industries.
Zertifizierungen: ANSI Cut A9. ANSI ABR 5. EN 388:2016 +A1:2018, EN407:2020, EN ISO 21420:2020. Food Contact approved. Category II.
Verpackung: Gloves are packaged 12 pairs per paper band bundle, 12 paper band bundles per carton.