Titan(III)oxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
481033-10G
481033-50G
SIAL481033-10GEA
72.6
EUR
SIAL481033-10G
SIAL481033-50G
Titan(III)oxid, Sigma-Aldrich®
Titan(III)oxid
Formel:
O₃Ti₂ Schmelzpunkt: 1900 °C Dichte: 4.49 g/cm³ (20 °C) |
MDL Number:
MFCD00036278 CAS-Nummer: 1344-54-3 EINECS: 215-697-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...