Coomassie Brillantblau G 250, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Brilliant blue G-250, Serva Blue G, Coomassie Blue G 250, Xylene Brilliant Cyanine G, Brillant blue G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, C.I. 42655, Acid Blue 90, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonate, Brilliant indocyanine G, Brilliant Blue G
Formel:
C₄₇H₄₈N₃NaO₇S₂ Molekulargewicht: 854.04 g/mol Dichte: Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00078482 CAS-Nummer: 6104-58-1 EINECS: 228-058-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...