N,N-Diethyl-m-toluamid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Diethyl-m-toluamide, Diethyltoluamide, N,N-Diethyl-3-methylbenzamide, Deet

A17486.0E A17486.22 A17486.36
A17486.0EEA 295 EUR
A17486.0E A17486.22 A17486.36
N,N-Diethyl-m-toluamid ≥97%
N,N-Diethyl-m-toluamid

Purity: 97% CAS No.: 134-62-3 Molecular Formula: C12H17NO Formula Weight: 191.27 Flash Point: 145°(293°F) Refractive Index: 1.5230 Boiling Point: 110-112°/1mm Beilstein Reference: 2046711 Merck Reference: 14,2856 MDL No.: MFCD00009046

Formel: C₁₂H₁₇NO
Siedepunkt: 110…112 °C (1 mmHg)
Schmelzpunkt: –45 °C
Dichte: 0.997
Flammpunkt: 145 °C (293 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00009046
CAS-Nummer: 134-62-3
EINECS: 205-149-7
Merck Index: 14,2856

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR