Primuline sodium salt

Lieferant: Thermo Fisher Scientific

473420010 473420050 473420250
473420010.EA 42.1 EUR
473420010. 473420050. 473420250.
Primuline sodium salt
Primuline sodium salt
Formel: C₂₁H₁₄N₃O₃S₃Na
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00005798
CAS-Nummer: 8064-60-6

Bestellen

Spezifikation Testergebnisse

Appearance yellow to brown powder or crystals
Infrared spectrum Conforms
UV Lambda max 350 to 370 nm (C=10,water)
Molar extinction coefficient ≥19000 (c=10, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR