2-Benzylbenzimidazole hydrochloride 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
473250010
473250050
473250010.EA
30.5
EUR
473250010.
473250050.
2-Benzylbenzimidazole hydrochloride 95%
2-Benzylbenzimidazole hydrochloride
Formel:
C₁₄H₁₂N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00443659 CAS-Nummer: 1212-48-2 EINECS: 214-921-2 |
Spezifikation Testergebnisse
Appearance | white to beige powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Titration with HClO4 | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...