EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)] 90%

Lieferant: Thermo Fisher Scientific

Synonyme: Bis(2,5-dioxopyrrolidin-1-yl) O,O'-ethane-1,2-diyl disuccinate, EGS, Ethylene glycol disuccinate di(N-succinimidyl) ester, Ethylene glycol bis(succinimidyl succinate), Di(N-succinimidyl) ethylene glycol disuccinate

472750010 472752500
472750010.EA 322 EUR
472750010. 472752500.
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)] 90%
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]
Formel: C₁₈H₂₀N₂O₁₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00049057
CAS-Nummer: 70539-42-3

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR