Cyclohexanesulphonyl chloride 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
472610010
472610050
472610010.EA
30.8
EUR
472610010.
472610050.
Cyclohexanesulphonyl chloride 95%
Cyclohexanesulphonyl chloride
Formel:
C₆H₁₁ClO₂S Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD04117775 CAS-Nummer: 4837-38-1 UN-Nummer: 3265 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | yellow liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...