N-Benzyl-9-(2-tetrahydropyranyl)adenine 97%
Lieferant: Thermo Fisher Scientific
472121000
472125000
472121000.EA
238
EUR
472121000.
472125000.
N-Benzyl-9-(2-tetrahydropyranyl)adenine 97%
N-Benzyl-9-(2-tetrahydropyranyl)adenine
Formel:
C₁₇H₁₉N₅O |
MDL Number:
MFCD00056971 CAS-Nummer: 2312-73-4 |
Spezifikation Testergebnisse
Appearance | white solid |
Infrared spectrum | Conforms |
HPLC | ≥96 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...