D-Galactono-1,4-lactone 95%
Lieferant: Thermo Fisher Scientific
471660010
471662500
471660010.EA
440
EUR
471660010.
471662500.
D-Galactono-1,4-lactone 95%
D-Galactono-1,4-lactone
Formel:
C₆H₁₀O₆ |
MDL Number:
MFCD00065019 CAS-Nummer: 2782-07-2 |
Spezifikation Testergebnisse
Appearance | white to light beige powder or crystals |
Infrared spectrum | Conforms |
Optical rotation | -70° to -80° (c=1 in water) |
NMR | ≥94 % Conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...