Bentonite, acid-leached (Catalyst 614-G)

Lieferant: Thermo Fisher Scientific

471230010 471230050 471231000
471230010.EA 54.7 EUR
471230010. 471230050. 471231000.
Bentonite, acid-leached (Catalyst 614-G)
Bentonite, acid-leached
Formel: H₂Al₂(SiO₃)₄-nH₂O
MDL Number: MFCD00132796
CAS-Nummer: 70131-50-9

Bestellen

Spezifikation Testergebnisse

Appearance beige to grey granules
Bulk density 475 to 625 kg/m³
Moisture 6 % to 10 %
Acidity ≤0.1 mg/KOH/g (Free Acidity)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR