Verbascoside 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Acteoside, 2-(3,4-Dihydroxyphenyl)ethyl 3-O-(6-Deoxy-alpha-L-mannopyranosyl)-4-O-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoyl]-beta-D-glucopyranoside, (2R,3R,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-5-hydroxy-2-(hydroxymethyl)-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl (E)-3-(3,4-dihydroxyphenyl)acrylate, Kusaginin
Formel:
C₂₉H₃₆O₁₅ Siedepunkt: 908.8°C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00221751 CAS-Nummer: 61276-17-3 |
Spezifikation Testergebnisse
Appearance | white to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...