Indole-3-butyric acid potassium salt 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Potassium 4-(1H-indol-3-yl)butanoate

471050010 471050050 471050250
471050010.EA 30.8 EUR
471050010. 471050050. 471050250.
Indole-3-butyric acid potassium salt 98%
Indole-3-butyric acid potassium salt
Formel: C₁₂H₁₂KNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00058442
CAS-Nummer: 60096-23-3

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow crystalline powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
Water ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR