(2-Aminophenyl)essigsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

471020010 471020050
471020010.EA 42.4 EUR
471020010. 471020050.
(2-Aminophenyl)essigsäure 98%
(2-Aminophenyl)essigsäure
Formel: C₈H₉NO₂
Siedepunkt: 344,4°C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03701521
CAS-Nummer: 3342-78-7

Bestellen

Spezifikation Testergebnisse

Appearance almost white to orange to light brown crystalline powder and or chunks
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 120°C to 127°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR