1,1,1-Tris(4-hydroxyphenyl)ethane 99%
Lieferant: Thermo Fisher Scientific
|
|
470940250
470941000
470940250.EA
30.5
EUR
470940250.
470941000.
1,1,1-Tris(4-hydroxyphenyl)ethane 99%
1,1,1-Tris(4-hydroxyphenyl)ethane
Formel:
C₂₀H₁₈O₃ Siedepunkt: 531,8°C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012180 CAS-Nummer: 27955-94-8 UN-Nummer: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | white to beige powder or crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Loss on drying | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...