3-Butynoic acid 97+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
But-3-ynoic acid
470870010
470870050
470870010.EA
82.9
EUR
470870010.
470870050.
3-Butynoic acid 97+%
3-Butynoic acid
Formel:
C₄H₄O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02258494 CAS-Nummer: 2345-51-9 UN-Nummer: 2923 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | white to almost white powder or crystals |
Infrared spectrum | Conforms |
Titration with NaOH | 97.0 to 103.0 % |
Melting point | 78.0°C to 87.0°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...