Mitoxantrone dihydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Novantrone, 1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione dihydrochloride
470790010
470792500
470790010.EA
288
EUR
470790010.
470792500.
Mitoxantrone dihydrochloride 97%
Mitoxantrone dihydrochloride
Formel:
C₂₂H₃₀Cl₂N₄O₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00242943 CAS-Nummer: 70476-82-3 EINECS: 274-619-1 UN-Nummer: 2811 ADR: 6.1,III Merck Index: 13,06238 |
Spezifikation Testergebnisse
Appearance | blue to black crystalline powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Water | ≤6 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...