Enzalutamide 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(3-(4-Cyano-3-(trifluoromethyl)phenyl)-5,5-dimethyl-4-oxo-2-thioxoimidazolidin-1-yl)-2-fluoro-N-methylbenzamide, 4-[3-[4-Cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-thioxo-imidazolidin-1-yl]-2-fluoro-N-methyl-benzamide, MDV-3100

470780010 470782500
470780010.EA 98.9 EUR
470780010. 470782500.
Enzalutamide 95%
Enzalutamide
Formel: C₂₁H₁₆F₄N₄O₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14155804
CAS-Nummer: 915087-33-1

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR