Ethyl-2-chlornicotinat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

470470050 470470250 470471000
470470050.EA 23.9 EUR
470470050. 470470250. 470471000.
Ethyl-2-chlornicotinat 98%
Ethyl-2-chlornicotinat
Formel: C₈H₈ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00674129
CAS-Nummer: 1452-94-4

Bestellen

Spezifikation Testergebnisse

Appearance clear colorless to light yellow liquid
Infrared spectrum Conforms
GC ≥97.5 %
Refractive index 1.5210 to 1.5250 (20°C, 589 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR