Di-tert-butyliminodiacetat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
470450010
470450050
470450010.EA
35.4
EUR
470450010.
470450050.
Di-tert-butyliminodiacetat 97%
Di-tert-butyl Iminodiacetate
Formel:
C₁₂H₂₃NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01863672 CAS-Nummer: 85916-13-8 |
Spezifikation Testergebnisse
Appearance | white to light brown crystalline powder or crystals |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Melting point | 38°C to 42°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...