3-(1H-Imidazol-4-yl)propanoic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Deamino-histidine, Imidazole-5-propionic acid, Imidazolepropionic acid

470380010 470382500
470380010.EA 250 EUR
470380010. 470382500.
3-(1H-Imidazol-4-yl)propanoic acid 97%
3-(1H-Imidazol-4-yl)propanoic acid
Formel: C₆H₈N₂O₂
Molekulargewicht: 140.142 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD18459271
CAS-Nummer: 1074-59-5
EINECS: 214-045-0

Bestellen

Spezifikation Testergebnisse

Appearance white to light grey solid
Infrared spectrum Conforms
HPLC ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR