Kaliumindol-3-acetat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

470280010 470280050 470280250
470280010.EA 25 EUR
470280010. 470280050. 470280250.
Kaliumindol-3-acetat 98%
Kaliumindol-3-acetat
Formel: C₁₀H₈KNO₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00066213
CAS-Nummer: 2338-19-4
EINECS: 219-049-6

Bestellen

Spezifikation Testergebnisse

Appearance white to orange to green crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR