Homoharringtonine 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Omacetaxine mepesuccinate
470100250
470101000
470100250.EA
262
EUR
470100250.
470101000.
Homoharringtonine 98%
Homoharringtonine
Formel:
C₂₉H₃₉NO₉ Molekulargewicht: 545,63 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD03410264 CAS-Nummer: 26833-87-4 UN-Nummer: 1544 ADR: 6.1 II |
Spezifikation Testergebnisse
Appearance | white to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 143°C to 148°C |
Water | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...