Sucralose 98%

Lieferant: Thermo Fisher Scientific

470090010 470091000 470092500
470090010.EA 165 EUR
470090010. 470091000. 470092500.
Sucralose 98%
Sucralose
Formel: C₁₂H₁₉Cl₃O₈
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03648615
CAS-Nummer: 56038-13-2
EINECS: 259-952-2
Merck Index: 13,08965

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +84° to +88° (20°C, 589 nm) (C=1% in water) (on dry substance)
Water ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR