Etretinate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ethyl all-trans-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoate, all-trans-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoic acid Ethyl ester, Acitretin ethyl ester

469960010 469962500
469960010.EA 286 EUR
469960010. 469962500.
Etretinate 98%
Etretinate
Formel: C₂₃H₃₀O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00866624
CAS-Nummer: 54350-48-0
EINECS: 259-119-3
UN-Nummer: 3077
ADR: 9,III
Merck Index: 13,03922

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to green crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 103°C to 107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR