Astragaloside IV 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2R,3R,4S,5S,6R)-2-(((2aR,3R,4S,5aS,5bS,7S,7aR,9S,11aR,12aS)-4-Hydroxy-3-((2R,5S)-5-(2-hydroxypropan-2-yl)-2-methyltetrahydrofuran-2-yl)-2a,5a,8,8-tetramethyl-9-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)tetradecahydro-1H,12H-cyclopenta[a]cyclopropa[e]phenanthren-7-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
468860010
468862500
468860010.EA
407
EUR
468860010.
468862500.
Astragaloside IV 98%
Astragaloside IV
Formel:
C₄₁H₆₈O₁₄ Molekulargewicht: 784.98 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD16036240 CAS-Nummer: 84687-43-4 |
Spezifikation Testergebnisse
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Water | ≤7 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...