Fenticonazole nitrate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
468790010
468792500
468790010.EA
236
EUR
468790010.
468792500.
Fenticonazole nitrate 98%
Fenticonazole nitrate
Formel:
C₂₄H₂₀Cl₂N₂OS·HNO₃ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00941391 CAS-Nummer: 73151-29-8 EINECS: 277-302-6 Merck Index: 13,04033 |
Spezifikation Testergebnisse
Appearance | white to almost white crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 133°C to 137°C |
Loss on drying | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...